Assay Design



Gene Transcripts

Gene Symbol
Pde1a
Entrez Gene ID
18573
Gene Name
phosphodiesterase 1A, calmodulin-dependent
Gene Aliases
AI987702, AW125737
Chromosome Location
Chr.2: 79834453 - 80196518 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009978.1 NP_001009978.1 8 - 9 887 99
NM_001009979.1 NP_001009979.1 8 - 9 887 99
NM_001159582.1 NP_001153054.1 10 - 11 1389 99
NM_016744.4 NP_058024.2 9 - 10 1184 99
GenBank mRNA AF159298.1 - 8 - 9 887 99
AK014810.1 - 8 - 9 98
AK043647.1 - 9 - 10 1185 99
AK050004.1 - 10 - 11 1251 99
AY845863.1 - 9 - 10 936 99
AY845864.1 - 8 - 9 887 99
BC090628.1 - 9 - 10 1152 99
U56649.1 - 10 - 11 1120 99

View Full Product Details