Assay Design



Gene Transcripts

Gene Symbol
Pigb
Entrez Gene ID
55981
Gene Name
phosphatidylinositol glycan anchor biosynthesis, class B
Gene Aliases
Pig-b
Chromosome Location
Chr.9: 73013726 - 73039699 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018889.3 NP_061377.2 1 - 2 238 78
GenBank mRNA AK153819.1 - 1 - 2 155 78
BC052658.1 - 1 - 2 198 78
D84436.1 - 1 - 2 238 78

View Full Product Details