Assay Design



Gene Transcripts

Gene Symbol
Tspan7
Entrez Gene ID
21912
Gene Name
tetraspanin 7
Gene Aliases
1200014P11Rik, A15, AI323365, Cd231, Mxs1, PE31, R74651, TALLA, Tm4sf2
Chromosome Location
Chr.X: 10485116 - 10596604 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019634.2 NP_062608.2 7 - 8 802 91
GenBank mRNA AF052492.1 - 7 - 8 750 91
AK004776.1 - 7 - 8 762 91
AK147306.1 - 6 - 7 5823 91
BC026516.1 - 6 - 7 601 91
BC050153.1 - 7 - 8 750 91
BC058603.1 - 7 - 8 758 91
D26483.1 - 7 - 8 750 91

View Full Product Details