Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019879.3 | NP_063932.2 | 7 - 8 | 974 | 77 |
GenBank mRNA | AF144101.2 | - | 7 - 8 | 807 | 77 |
AK002754.1 | - | 7 - 8 | 809 | 77 | |
AK005080.1 | - | 7 - 8 | 823 | 77 | |
AK088888.1 | - | 7 - 8 | 831 | 77 | |
BC011087.1 | - | 7 - 8 | 795 | 77 |