Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022415.3 | NP_071860.1 | 2 - 3 | 293 | 87 |
GenBank mRNA | AB035323.1 | - | 2 - 3 | 245 | 87 |
AB041997.1 | - | 2 - 3 | 258 | 87 | |
AK010757.1 | - | 2 - 3 | 258 | 87 | |
AK033752.1 | - | 2 - 3 | 258 | 87 | |
AK080897.1 | - | 2 - 3 | 298 | 87 | |
AK167015.1 | - | 2 - 3 | 162 | 87 | |
AK172621.1 | - | 2 - 3 | 258 | 87 | |
BC024960.1 | - | 2 - 3 | 252 | 87 |