Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022653.4 | NP_073144.3 | 1 - 2 | 197 | 94 |
GenBank mRNA | AF314187.1 | - | 1 - 2 | 155 | 94 |
AK028609.1 | - | 1 - 2 | 131 | 94 | |
BC003211.1 | - | 1 - 2 | 153 | 94 | |
BC031722.1 | - | 1 - 2 | 163 | 94 | |
CF163503.1 | - | 1 - 2 | 178 | 94 |