Assay Design



Gene Transcripts

Gene Symbol
Palm
Entrez Gene ID
18483
Gene Name
paralemmin
Gene Aliases
-
Chromosome Location
Chr.10: 79793351 - 79820896 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023128.4 NP_075617.3 8 - 9 794 78
GenBank mRNA AK037044.1 - 8 - 9 651 78
AK129107.1 - 6 - 7 533 78
Y14771.1 - 8 - 9 757 78

View Full Product Details