Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026835.2 | NP_081111.1 | 6 - 7 | 797 | 119 |
GenBank mRNA | AF237911.1 | - | 6 - 7 | 759 | 119 |
AK004295.1 | - | 6 - 7 | 780 | 119 | |
AK152859.1 | - | 6 - 7 | 778 | 119 | |
AK156878.1 | - | 6 - 7 | 780 | 119 | |
AY039037.1 | - | 6 - 7 | 759 | 119 | |
BC018331.1 | - | 6 - 7 | 734 | 119 | |
BC029738.1 | - | 6 - 7 | 741 | 119 |