Assay Design



Gene Transcripts

Gene Symbol
Osbpl3
Entrez Gene ID
71720
Gene Name
oxysterol binding protein-like 3
Gene Aliases
1200014M06Rik, 6720421I08Rik, A530055M08, ORP3, OSBP3
Chromosome Location
Chr.6: 50293326 - 50456355 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163645.1 NP_001157117.1 6 - 7 957 67
NM_027881.3 NP_082157.2 5 - 6 567 67
GenBank mRNA AK004768.1 - 5 - 6 569 67
AK037114.1 - 5 - 6 567 67
AK173006.1 - 6 - 7 766 67
BC044866.1 - 5 - 6 602 67
BC086485.1 - 6 - 7 869 67

View Full Product Details