Assay Design
Gene Transcripts
- Gene Symbol
- Sall4
- Entrez Gene ID
-
99377
- Gene Name
- sal-like 4 (Drosophila)
- Gene Aliases
-
5730441M18Rik, AA407717, AL022809, AW536104, C330011P20Rik, C78083, C78563, Tex20
- Chromosome Location
-
Chr.2: 168748332 - 168767201 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF285588.1 |
- |
1 - 1 |
448 |
93 |
AK017633.1 |
- |
1 - 1 |
948 |
93 |
AK145388.1 |
- |
4 - 4 |
4327 |
93 |
BC067396.1 |
- |
1 - 1 |
750 |
93 |
View Full Product Details