Assay Design



Gene Transcripts

Gene Symbol
Trim2
Entrez Gene ID
80890
Gene Name
tripartite motif-containing 2
Gene Aliases
mKIAA0517, narf
Chromosome Location
Chr.3: 84160439 - 84305832 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030706.3 NP_109631.2 1 - 2 252 91
GenBank mRNA AB043550.1 - 1 - 2 236 91
AK039741.1 - 1 - 2 250 91
AK045410.1 - 1 - 2 249 91
AK046887.1 - 1 - 2 237 91
AK083057.1 - 1 - 2 250 91
AK147395.1 - 1 - 2 247 91
BC058668.1 - 1 - 2 220 91
BY124765.1 - 1 - 2 262 91
CD803806.1 - 1 - 2 252 91

View Full Product Details