Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282014.1 | NP_001268943.1 | 7 - 8 | 1678 | 123 |
NM_001282015.1 | NP_001268944.1 | 7 - 8 | 1672 | 123 | |
NM_031202.3 | NP_112479.1 | 7 - 8 | 1693 | 123 | |
GenBank mRNA | AK148332.1 | - | 7 - 8 | 1678 | 123 |
AK181989.1 | - | 2 - 3 | 177 | 123 | |
AK188091.1 | - | 2 - 3 | 179 | 123 | |
AK193585.1 | - | 2 - 3 | 179 | 123 | |
AK194427.1 | - | 2 - 3 | 179 | 123 | |
AK194794.1 | - | 1 - 2 | 156 | 123 | |
AK194981.1 | - | 1 - 2 | 159 | 123 | |
AK198402.1 | - | 1 - 2 | 169 | 124 | |
AK199163.1 | - | 2 - 3 | 179 | 123 | |
AK199872.1 | - | 2 - 3 | 179 | 123 | |
AK200013.1 | - | 2 - 3 | 179 | 123 | |
AK200365.1 | - | 2 - 3 | 179 | 123 | |
AK203562.1 | - | 1 - 2 | 134 | 123 | |
AK207921.1 | - | 2 - 3 | 192 | 123 | |
BC076598.1 | - | 7 - 8 | 1676 | 123 | |
X03687.1 | - | 7 - 8 | 1578 | 123 |