Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159671.1 | NP_001153143.1 | 3 - 4 | 1063 | 61 |
NM_031255.2 | NP_112545.2 | 4 - 5 | 1828 | 61 | |
GenBank mRNA | AF329191.1 | - | 3 - 4 | 1051 | 61 |
AF329192.1 | - | 4 - 5 | 1821 | 61 | |
AK029699.1 | - | 4 - 5 | 1828 | 61 | |
BC137992.1 | - | 4 - 5 | 1821 | 61 |