Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163776.1 | NP_001157248.1 | 2 - 3 | 295 | 102 |
NM_080727.2 | NP_542765.2 | 1 - 2 | 155 | 102 | |
GenBank mRNA | AJ300738.1 | - | 1 - 2 | 155 | 102 |
AK158203.1 | - | 2 - 3 | 298 | 102 | |
BC112375.1 | - | 1 - 2 | 152 | 102 |