Assay Design



Gene Transcripts

Gene Symbol
Ptov1
Entrez Gene ID
84113
Gene Name
prostate tumor over expressed gene 1
Gene Aliases
1110030G05Rik, AU041779, Gcap3
Chromosome Location
Chr.7: 44863067 - 44870411 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133949.1 NP_598710.1 7 - 8 1068 66
GenBank mRNA BC008543.1 - 7 - 8 944 66
BC024632.1 - 7 - 8 1068 66

View Full Product Details