Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138676.2 | NP_619617.2 | 9 - 10 | 894 | 91 |
GenBank mRNA | AF246218.1 | - | 9 - 10 | 900 | 91 |
AK047434.1 | - | 7 - 8 | 1025 | 91 | |
AK154779.1 | - | 9 - 10 | 883 | 90 | |
AK160641.1 | - | 8 - 9 | 966 | 91 | |
BC061477.1 | - | 9 - 10 | 847 | 91 |