Assay Design



Gene Transcripts

Gene Symbol
Rfpl4
Entrez Gene ID
192658
Gene Name
ret finger protein-like 4
Gene Aliases
D7Ertd486e, Rfpl4a
Chromosome Location
Chr.7: 5109787 - 5116911 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138954.3 NP_620404.1 1 - 2 277 86
GenBank mRNA AK139569.1 - 1 - 2 279 86
AY070253.1 - 1 - 2 267 86

View Full Product Details