Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139140.1 | NP_631879.1 | 12 - 13 | 1640 | 77 |
GenBank mRNA | AB079132.1 | - | 12 - 13 | 1640 | 77 |
AK159335.1 | - | 12 - 13 | 1735 | 77 | |
AK167788.1 | - | 11 - 12 | 1318 | 77 | |
AK171266.1 | - | 11 - 12 | 1396 | 77 | |
BC023432.1 | - | 10 - 11 | 1290 | 77 | |
BC072579.1 | - | 12 - 13 | 1907 | 77 |