Assay Design



Gene Transcripts

Gene Symbol
Mcfd2
Entrez Gene ID
193813
Gene Name
multiple coagulation factor deficiency 2
Gene Aliases
1810021C21Rik, F5f8d, Lman1ip, Sdnsf
Chromosome Location
Chr.17: 87254443 - 87265947 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139295.3 NP_647456.1 2 - 3 272 63
NM_176808.5 NP_789778.1 3 - 4 497 63
GenBank mRNA AF475283.1 - 2 - 3 240 63
AK033139.1 - 2 - 3 206 63
AK075781.1 - 3 - 4 430 63
AK077139.1 - 2 - 3 223 63
AK077631.1 - 2 - 3 206 63
AK141599.1 - 3 - 4 455 63
AK151803.1 - 2 - 3 261 63
AK206943.1 - 1 - 2 151 63
BC003996.1 - 2 - 3 199 63
GQ896355.1 - 1 - 2 143 63
GQ896356.1 - 1 - 2 143 63

View Full Product Details