Assay Design



Gene Transcripts

Gene Symbol
Spata20
Entrez Gene ID
217116
Gene Name
spermatogenesis associated 20
Gene Aliases
Tisp78
Chromosome Location
Chr.11: 94478904 - 94485310 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144827.4 NP_659076.2 15 - 16 2230 77
GenBank mRNA AB045721.2 - 1 - 2 77
BC050788.1 - 15 - 16 2230 77

View Full Product Details