Assay Design



Gene Transcripts

Gene Symbol
Olfr30
Entrez Gene ID
18329
Gene Name
olfactory receptor 30
Gene Aliases
MOR2811, MTPCR07
Chromosome Location
Chr.11: 58454915 - 58455980 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146878.2 NP_667089.1 1 - 1 480 64
GenBank mRNA AY043304.1 - 1 - 1 64 64
BC119375.1 - 1 - 1 480 64
X89680.1 - 1 - 1 70 64

View Full Product Details