Assay Design



Gene Transcripts

Gene Symbol
Rps6ka3
Entrez Gene ID
110651
Gene Name
ribosomal protein S6 kinase polypeptide 3
Gene Aliases
C130006E23, MAPKAPK-1b, MPK-9, Rsk2, S6K-alpha3, p90RSK3, pp90RSK2
Chromosome Location
Chr.X: 159255782 - 159368244 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_148945.2 NP_683747.1 3 - 4 577 108
GenBank mRNA AK079102.1 - - 670 108
AK154778.1 - 3 - 4 490 108
AY083469.1 - 3 - 4 239 108

View Full Product Details