Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152800.3 | NP_690013.1 | 1 - 2 | 215 | 62 |
GenBank mRNA | AK162719.1 | - | 1 - 2 | 159 | 62 |
AK171247.1 | - | 1 - 2 | 167 | 62 | |
BC003466.1 | - | 1 - 2 | 200 | 62 | |
BC024469.1 | - | 1 - 2 | 178 | 62 |