Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271616.1 | NP_001258545.1 | 3 - 3 | 900 | 66 |
NM_008009.4 | NP_032035.2 | 2 - 2 | 850 | 66 | |
GenBank mRNA | AK146625.1 | - | 2 - 2 | 815 | 66 |
BC065774.1 | - | 2 - 2 | 813 | 66 | |
CB524251.1 | - | 1 - 1 | 160 | 66 | |
U49641.1 | - | 1 - 1 | 1848 | 66 |