Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008576.3 | NP_032602.1 | 5 - 6 | 653 | 67 |
GenBank mRNA | AF022908.1 | - | 5 - 6 | 615 | 67 |
AK029876.1 | - | 5 - 6 | 654 | 67 | |
AK150015.1 | - | 5 - 6 | 634 | 67 | |
AK152781.1 | - | 5 - 6 | 634 | 67 | |
BC090617.1 | - | 5 - 6 | 624 | 67 |