Assay Design



Gene Transcripts

Gene Symbol
Abcc1
Entrez Gene ID
17250
Gene Name
ATP-binding cassette, sub-family C (CFTR/MRP), member 1
Gene Aliases
Abcc1a, Abcc1b, MRP, Mdrap, Mrp1
Chromosome Location
Chr.16: 14361521 - 14474879 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008576.3 NP_032602.1 20 - 21 2768 60
GenBank mRNA AF022908.1 - 20 - 21 2730 60
AK029876.1 - 20 - 21 2769 60
AK049174.1 - 4 - 5 535 60
BC090617.1 - 20 - 21 2739 60

View Full Product Details