Assay Design



Gene Transcripts

Gene Symbol
Dusp8
Entrez Gene ID
18218
Gene Name
dual specificity phosphatase 8
Gene Aliases
5530400B01Rik, AI593498, Nttp1
Chromosome Location
Chr.7: 142079487 - 142095880 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008748.3 NP_032774.1 2 - 3 688 82
GenBank mRNA BC052705.1 - 2 - 3 446 82
X95518.1 - 1 - 2 331 82

View Full Product Details