Assay Design



Gene Transcripts

Gene Symbol
Stmn3
Entrez Gene ID
20262
Gene Name
stathmin-like 3
Gene Aliases
9330161A03Rik, AI604839, R75323, Sclip
Chromosome Location
Chr.2: 181306459 - 181314500 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009133.3 NP_033159.1 1 - 2 301 122
GenBank mRNA AB007912.1 - 1 - 2 89 122
AK032737.1 - 1 - 2 302 122

View Full Product Details