Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009212.2 | NP_033238.2 | 10 - 11 | 1756 | 93 |
GenBank mRNA | AK142456.1 | - | 10 - 11 | 1560 | 93 |
AK153763.1 | - | 10 - 11 | 1717 | 93 | |
BC052025.1 | - | 9 - 10 | 1628 | 93 | |
BC060085.1 | - | 9 - 10 | 1628 | 93 | |
BC066215.1 | - | 10 - 11 | 1699 | 93 | |
BC083142.1 | - | 10 - 11 | 1561 | 93 | |
L10075.1 | - | 10 - 11 | 1652 | 93 |