Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010826.5 | NP_034956.2 | 2 - 3 | 454 | 76 |
NM_194464.3 | NP_919446.2 | 3 - 4 | 697 | 76 | |
GenBank mRNA | AK020536.1 | - | 3 - 4 | 279 | 76 |
U63407.1 | - | 3 - 4 | 700 | 76 | |
U63408.1 | - | 2 - 3 | 449 | 76 |