Assay Design



Gene Transcripts

Gene Symbol
Mrvi1
Entrez Gene ID
17540
Gene Name
MRV integration site 1
Gene Aliases
BB115629, Irag, Ris1
Chromosome Location
Chr.7: 110868257 - 110982908 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010826.5 NP_034956.2 13 - 14 1996 65
NM_194464.3 NP_919446.2 14 - 15 2239 65
GenBank mRNA AK141821.1 - 9 - 10 1278 65
AK157169.1 - 14 - 15 1849 65
U63407.1 - 14 - 15 2242 65
U63408.1 - 13 - 14 1991 65

View Full Product Details