Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029843.1 | NP_001025014.1 | 4 - 5 | 387 | 90 |
NM_001029844.1 | NP_001025015.1 | 4 - 5 | 387 | 90 | |
NM_011705.3 | NP_035835.1 | 4 - 5 | 387 | 90 | |
GenBank mRNA | AF080252.1 | - | 3 - 4 | 300 | 90 |
AF080253.1 | - | 4 - 5 | 341 | 90 | |
AK032344.1 | - | 4 - 5 | 358 | 90 | |
AK088230.1 | - | 4 - 5 | 317 | 90 | |
AK088674.1 | - | 4 - 5 | 370 | 90 | |
AK136352.1 | - | 4 - 5 | 361 | 90 | |
AK166676.1 | - | 4 - 5 | 355 | 90 | |
BC016676.1 | - | 4 - 5 | 422 | 90 | |
BC051072.1 | - | 4 - 5 | 367 | 90 |