Assay Design



Gene Transcripts

Gene Symbol
Zpbp
Entrez Gene ID
53604
Gene Name
zona pellucida binding protein
Gene Aliases
4930486K01Rik, Iam38, Sp38
Chromosome Location
Chr.11: 11279624 - 11463630 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001185153.1 NP_001172082.1 5 - 6 762 99
NM_015785.2 NP_056600.1 5 - 6 933 99
GenBank mRNA AK006274.1 - 2 - 3 275 99
AK018889.1 - 2 - 3 277 99
AK018921.1 - 2 - 3 277 99
AK029541.1 - 2 - 3 277 99
BC139450.1 - 5 - 6 902 99
BC139453.1 - 5 - 6 902 99
BC145628.1 - 5 - 6 731 99
D17569.1 - 5 - 6 922 99

View Full Product Details