Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017371.2 | NP_059067.2 | 9 - 10 | 1148 | 63 |
GenBank mRNA | AA985967.1 | - | 1 - 2 | 63 | |
AK145928.1 | - | 9 - 10 | 1146 | 63 | |
BC011246.1 | - | 9 - 10 | 1128 | 63 | |
BC019901.1 | - | 9 - 10 | 1130 | 63 | |
U89889.1 | - | 9 - 10 | 1104 | 63 |