Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023603.3 | NP_076092.1 | 4 - 5 | 1511 | 66 |
NR_045010.1 | 4 - 5 | 1511 | 66 | ||
GenBank mRNA | AF110499.1 | - | 2 - 3 | 293 | 66 |
AF272847.1 | - | 4 - 5 | 804 | 66 | |
AK179158.1 | - | 2 - 3 | 199 | 66 | |
AK210812.1 | - | 2 - 3 | 123 | 66 | |
AK216272.1 | - | 2 - 3 | 115 | 66 | |
AY034062.1 | - | 4 - 5 | 1426 | 66 | |
BC037486.1 | - | 2 - 3 | 224 | 66 | |
BC049227.1 | - | 1 - 2 | 82 | 66 | |
BC080848.1 | - | 4 - 5 | 1477 | 66 | |
BC089305.1 | - | 4 - 5 | 1477 | 66 | |
CA319647.1 | - | 2 - 3 | 142 | 66 |