Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023637.3 | NP_076126.2 | 14 - 15 | 1393 | 108 |
GenBank mRNA | AB029949.1 | - | 14 - 15 | 1365 | 108 |
AK010491.1 | - | 14 - 15 | 1371 | 108 | |
AK087606.1 | - | 14 - 15 | 1360 | 108 | |
AK140760.1 | - | 14 - 15 | 1360 | 108 | |
BC079664.1 | - | 14 - 15 | 1370 | 108 |