Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026206.2 | NP_080482.1 | 1 - 2 | 89 | 76 |
GenBank mRNA | AF525157.1 | - | 1 - 2 | 74 | 76 |
AK005486.1 | - | 1 - 2 | 90 | 76 | |
AK014436.1 | - | 1 - 2 | 73 | 76 | |
BC099442.1 | - | 1 - 2 | 49 | 76 |