Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026370.1 | NP_080646.1 | 6 - 7 | 780 | 91 |
GenBank mRNA | AK003264.1 | - | 6 - 7 | 780 | 91 |
AK008418.1 | - | 6 - 7 | 767 | 91 | |
AK018002.1 | - | 6 - 7 | 776 | 91 | |
AK132000.1 | - | 6 - 7 | 781 | 91 | |
AK146710.1 | - | 6 - 7 | 782 | 91 | |
BC036284.1 | - | 6 - 7 | 788 | 91 |