Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027022.4 | NP_081298.1 | 2 - 3 | 446 | 79 |
GenBank mRNA | AK005603.1 | - | 2 - 3 | 399 | 79 |
AK161303.1 | - | 2 - 3 | 440 | 79 | |
AY172740.1 | - | 2 - 3 | 327 | 79 | |
AY241871.1 | - | 2 - 3 | 446 | 79 | |
BC049747.1 | - | 2 - 3 | 439 | 79 |