Assay Design



Gene Transcripts

Gene Symbol
Actr8
Entrez Gene ID
56249
Gene Name
ARP8 actin-related protein 8
Gene Aliases
5730542K05Rik, ARP8
Chromosome Location
Chr.14: 29978330 - 29993221 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027493.3 NP_081769.2 4 - 5 639 131
GenBank mRNA AK017815.1 - 4 - 5 590 131
BC027281.1 - 4 - 5 545 131

View Full Product Details