Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028207.3 | NP_082483.1 | 1 - 2 | 182 | 59 |
GenBank mRNA | AF280809.1 | - | 1 - 2 | 147 | 59 |
AK008734.1 | - | 1 - 2 | 163 | 59 | |
AK170772.1 | - | 1 - 2 | 163 | 59 | |
BC016269.1 | - | 1 - 2 | 190 | 59 |