Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030691.1 | NP_109616.1 | 1 - 2 | 94 | 76 |
GenBank mRNA | AJ278575.1 | - | 1 - 2 | 94 | 76 |
AK151417.1 | - | 1 - 2 | 83 | 76 | |
AK152186.1 | - | 1 - 2 | 83 | 76 | |
AK163058.1 | - | 1 - 2 | 74 | 76 | |
BC057951.1 | - | 1 - 2 | 91 | 76 |