Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031381.2 | NP_113558.1 | 3 - 4 | 710 | 81 |
GenBank mRNA | AF285576.1 | - | 3 - 4 | 710 | 81 |
AK016637.1 | - | 1 - 2 | 848 | 81 | |
BC116863.1 | - | 3 - 4 | 686 | 81 | |
BC119125.1 | - | 3 - 4 | 686 | 81 |