Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033572.2 | NP_291050.1 | 1 - 2 | 368 | 93 |
GenBank mRNA | AF410456.1 | - | 1 - 2 | 347 | 93 |
AK005040.1 | - | 1 - 2 | 314 | 93 | |
AK017733.1 | - | 1 - 2 | 367 | 93 | |
BC024714.1 | - | 1 - 2 | 340 | 93 | |
BC055335.1 | - | 1 - 2 | 366 | 93 | |
BY725117.1 | - | 1 - 2 | 376 | 93 |