Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033572.2 | NP_291050.1 | 6 - 7 | 833 | 98 |
GenBank mRNA | AF410456.1 | - | 6 - 7 | 812 | 98 |
AK005040.1 | - | 6 - 7 | 779 | 98 | |
AK017733.1 | - | 6 - 7 | 832 | 98 | |
AK177410.1 | - | 3 - 4 | 169 | 99 | |
BC024714.1 | - | 6 - 7 | 805 | 98 | |
BC055335.1 | - | 6 - 7 | 831 | 98 |