Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110827.2 | NP_001104297.1 | 9 - 10 | 1475 | 67 |
NM_001110829.2 | NP_001104299.1 | 8 - 9 | 1017 | 67 | |
NM_001110830.2 | NP_001104300.1 | 8 - 9 | 1017 | 67 | |
NM_001286417.1 | NP_001273346.1 | 8 - 9 | 1017 | 67 | |
NM_001286419.1 | NP_001273348.1 | 7 - 8 | 921 | 67 | |
NM_175387.3 | NP_780596.1 | 9 - 10 | 1475 | 67 | |
GenBank mRNA | AF229055.1 | - | 8 - 9 | 960 | 67 |
AF387322.1 | - | 8 - 9 | 761 | 67 | |
AK044929.1 | - | 8 - 9 | 992 | 67 | |
AY659951.1 | - | 8 - 9 | 761 | 67 | |
AY659952.1 | - | 8 - 9 | 761 | 67 | |
AY904025.1 | - | 10 - 11 | 911 | 67 | |
BC002124.1 | - | 8 - 9 | 855 | 67 | |
BC027263.1 | - | 9 - 10 | 1027 | 67 | |
DQ017389.1 | - | 7 - 8 | 665 | 67 |