Assay Design



Gene Transcripts

Gene Symbol
Ms4a3
Entrez Gene ID
170813
Gene Name
membrane-spanning 4-domains, subfamily A, member 3
Gene Aliases
HTm4
Chromosome Location
Chr.19: 11629499 - 11640851 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133246.5 NP_573509.1 5 - 6 652 63
GenBank mRNA AF321128.1 - 5 - 6 591 63
AK036464.1 - 5 - 6 654 63
AY184359.1 - 5 - 6 606 63
BC117013.1 - 5 - 6 568 63
BC117015.1 - 5 - 6 568 63
BC144853.1 - 5 - 6 568 63

View Full Product Details