Assay Design



Gene Transcripts

Gene Symbol
Msl3l2
Entrez Gene ID
73390
Gene Name
male-specific lethal 3-like 2 (Drosophila)
Gene Aliases
1700060H10Rik
Chromosome Location
Chr.10: 56083827 - 56116880 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163833.1 NP_001157305.1 1 - 2 110 102
GenBank mRNA AK155382.1 - 1 - 2 96 102
BC037487.1 - 1 - 2 121 102

View Full Product Details