Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133779.2 | NP_598540.2 | 2 - 3 | 413 | 77 |
GenBank mRNA | AK019717.2 | - | 2 - 3 | 404 | 77 |
AK044499.1 | - | 2 - 3 | 397 | 77 | |
AK049171.1 | - | 2 - 3 | 407 | 77 | |
AK049990.1 | - | 2 - 3 | 406 | 77 | |
AK090020.1 | - | 2 - 3 | 406 | 77 | |
AK153462.1 | - | 2 - 3 | 399 | 77 | |
AK153464.1 | - | 2 - 3 | 399 | 77 | |
AK155057.1 | - | 2 - 3 | 400 | 77 | |
AK157226.1 | - | 2 - 3 | 396 | 77 | |
AK162573.1 | - | 2 - 3 | 393 | 77 | |
BC034175.1 | - | 2 - 3 | 389 | 77 | |
BU058874.1 | - | 2 - 3 | 413 | 77 |