Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113401.1 | NP_001106872.1 | 5 - 6 | 844 | 69 |
NM_001113405.1 | NP_001106876.1 | 3 - 4 | 486 | 69 | |
NM_134111.2 | NP_598872.1 | 5 - 6 | 834 | 69 | |
GenBank mRNA | AB081298.1 | - | 3 - 4 | 486 | 69 |
AK076871.1 | - | 6 - 7 | 781 | 69 | |
AY034479.1 | - | 5 - 6 | 851 | 69 | |
AY049021.1 | - | 5 - 6 | 790 | 69 | |
BC004721.1 | - | 5 - 6 | 833 | 69 | |
BC056626.1 | - | 5 - 6 | 834 | 69 |